Painted human skull model, 3 parts
Catalog # BINH1000055
Supplier: 3B Scientific
Specifications
- Poids:0,7 kg
- Désignation:Painted human skull model
- Cdt:1
Specifications
About this item
The muscle origins (red) and insertions (blue) are shown in color on the left side of the skull. Cranial bones and structures are numbered on the right side. This skull model identifies over 140 anatomical details:
- High-quality original casts
- Handmade of hard, unbreakable plastic
- Highly accurate representation of the fissures, foramina, processes, sutures
- Can be disassembled into skull cap, base of skull and mandible
- As an option, you can insert a five-part brain (C 18) into the skull
This replica of the human skull is designed for teaching and learning the anatomy of the skull.