Human skull model on cervical spine, 4 part
BINH1000047
- Désignation:Human skull model on cervical spine, 4 part
- Cdt:1
This human skull replica is flexibly mounted on a cervical spine. Features also shown are the hindbrain, spinal cord, cervical nerves, vertebral arteries, basilar artery and rear cerebral arteries. Skull can be removed from stand.
- High quality original human skull cast
- Skull is handmade from hard, unbreakable plastic
- Highly accurate representation of the fissures, foramina, processes, sutures etc.
- Can be disassembled into skull cap, base of skull and mandible
- Mandible of skull is mounted on a spring to easily demonstrate natural movement
Dimensions: 20×13,5×15,5 cm
Weight: 0,6 kg