Bouffant dispenser with lid
SCURBD-600
: S-CURVE
- Couleur:incolore
- Désignation:Dispenser for bouffant caps, with hinged lid and front large round opening, 1 compartment, acrylic
- l×p×h:304,8×203,2×304,8 mm
- Cdt:1
Bouffant dispenser, made of clear acrylic, for cleanroom bouffants.
- Single compartment
- Lid with full length piano hinge
- Large round opening for easy access
- Flame treated edges for a smooth finish
- Keyholes for wall mounting