Weighing boats
N° de catalogue DELT1911103
Fournisseur: DELTALAB
Spécifications
- l×p×h:41×41×8 mm
- Cdt:500
Spécifications
À propos de cet article
PS, square, white.
- Resistant to weak acids, water based solutions and alcohol
- Can be folded for easy pouring
- Suitable for microwave