Pinces pour tuyaux
Catalog # 229-0374
Supplier: HAMMACHER KARL
Specifications
- Longueur:200 mm
- Seq:01
- Cdt:1
Specifications
About this item
Tubing clamps made of stainless steel.
- Fingers are covered with plastic coating to protect clamped items
- Corrosion resistant