Iode concentré pour une dilution à 1L, 0.05 M I2 (0.1 N) solution volumétrique, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
EINECS:231-442-4
:GHS07
:GHS08
Storage Temperature:Température ambiante
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
Frequently Bought Together