Racks for EasyLoad® pipette tips
Référence GREI941310_10
Fournisseur: Greiner Bio-One
Spécifications
- Code couleur:jaune
- Désignation:Boîte pour portoir de pointes EasyLoad® 200
- Cdt:10
Spécifications
À propos de cet article
EasyLoad® is a simple, time- and space-saving refill system for pipette tips. The refill system also contains EasyLoad® racks specially developed for this purpose.
- Autoclavable EasyLoad® racks
- Sterile and non-sterile option
- Racks are stackable
- Colour-coded boxes
The modularly constructed EasyLoad® racks are available in different sizes. They consist of a polypropylene box with attached lid and a polypropylene insert plate. Users of the EasyLoad® refill system will find the insert plate included in the refill unit. The EasyLoad® racks are also particularly well suited for use with multi-channel pipettors, since the individual tips in the rack can be picked up with the same force.