Storage boxes, with lid, neoLab®
Fournisseur: NEOLAB
Certificats
À propos de cet article
These stackable boxes with lid made of polycarbonate (PC) are ideal for the storage of the LaboBox® racks.
- Higher boxes accepts microscopy slides of 76×45 mm
- Can be used in cryogenic applications
- Temperature resistant from −130 to +130 °C
Frequently Bought Together