Protective hand creams, Sensiva®
Catalog # SCUE116001
Supplier: SCHÜLKE & MAYR GMBH
Specifications
- Désignation:Protective hand cream, 150 ml flask
- Seq:01
- Cdt:30
Specifications
About this item
Sensiva® Protective Emulsion O/W protects and maintains the moisture of normal and combination skin.
- Oil-in-water emulsion
- Free of colourant and perfume
- Absorbs quickly, protects and nourishes the skin
- Does not reduce the effect of hand disinfectant
Caution: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904