Visual comparator test kit for dssolved oxygen by the indiigo carmine method, CHEMets
Catalog # CHETK-7512
Supplier: CHEMetrics
CAS Number:
Specifications
- Cdt:1 KIT
- Désignation:Dissolved oxygen kit, indigo carmne method
- Plage de mesure:1-2-3-4-5-6-8-10-12 ppm
Specifications
About this item
Test kits for environmental and drinking water applications in the ppm range.
- Visual indication of dissolved oxygen by formation of blue product
- Method is not to interferences from temperature, salinity, or dissolved gases such as sulfide
- Results are expressed as ppm (mg/L) O₂
Delivery: Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, comparator, 25 ml sample cup and instructions