Dismofix® N Neutral Detergent
Catalog # BODL972940
Supplier: Bode
Specifications
- Désignation:Dismofix® N, neutral detergent for washable surfaces, 5-litre canister
- Cdt:12
Specifications
About this item
This multipurpose neutral detergent is particularly suitable for thorough and sparing neutral cleaning of all washable surfaces and objects, but also for cleaning in combination with all surface disinfectants from BODE. The detergent is highly concentrated (and therefore particularly economical), pH neutral and fully bio-degradable.
Composition: non-ionic surfactants, solubilisers, sequestrants, scents.
Registered as a tested detergent for ceramic surfaces in swimming pools (Robert Koch Institute list).
Caution: Suggestion: Before applying on coated floors, it is advisable to test the consistency in an inconspicuous place.