A-1 medium broth MPN tubes
HACH2560915
: Hach
:
- Cdt:15
- Normes:-
- Désignation:A-1 medium broth MPN tubes
A-1 Medium broth tubes provide detection of faecal coliform bacteria using the Most Probable Number (MPN) method.
- Save time and labor costs
- Minimal training requirements