Hand cleanser dispenser, Combiplum
115-0783
- Capacité:1000 ml
- Couleur:blanc
- Désignation:Electronic touch free dispenser
- l×p×h:140×100×307 mm
- Cdt:1
This electronic touch free dispenser for 1,0 L CombiPlum bags is designed for all working areas with particularly high hygiene standards.
: Supplied with four AA batteries (included).