Screw thread vials
Catalog # NSCAC4015-S11W
Supplier: THERMO NATIONAL SCIENTIFIC
Specifications
- Avec bouchon:Non
- Ø×h:15×45 mm
- Capacité:4,00 ml
- Couleur:incolore
- Désignation:Silanised I-D™ vials, with write-on patch
- Matière:Verre
- Seq:06
- Fond:Flat bottom
- Cdt:100
Specifications
About this item
Original Waters™ 4 ml WISP™ autosampler vials. Superior quality 33 expansion borosilicate clear glass (Type 1, Class A) or 51A amber glass.