Cylinders, mutipurpose
200 mm, 40 mm
N° de catalogue WITG5.630.004
Fournisseur: witeg Labortechnik
Aller à
Product Details & Documents
Cylinders, made of DURAN® borosilicate glass, with rolled rim and round base.
- Very good chemical resistance
- High resistance to temperature change
- No graduations
Spécifications
- Ø:40 mm
- Hauteur:200 mm