HEPES (Acide 4-(2-hydroxyéthyl)pipérazine-1-yléthanesulfonique)
Catalog # GRBU1009.2500
Supplier: GERBU BIOCHEMICALS
CAS Number:
Specifications
- Type de conditionnement:Glass bottle for solids
- Cdt:2.500 g
- Seq:3795
Specifications
About this item
:
Merck Index:13,04672
Melting Pt:210…215 °C
Density:1,448 g/cm³ (20 °C)
:238.31 g/mol
Storage Temperature:Température ambiante
EINECS:230-907-9
CAS Number:7365-45-9
Formula:C₈H₁₈N₂O₄S
MDL Number:MFCD00006158