Chromium standard dissolved in oil, 5,000 µg/g Cr in mineral oil Specpure® for wear metal analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
Physical Form: Liquid
MDL No.: MFCD00010944
Specifications
- MDL No.:MFCD00010944
Specifications
Related Information
Frequently Bought Together