ContactUsLinkComponent
Refrigerated Heating Circulator Temperature range: -60 ... 200 °C Cooling capacity (0°C): 7 kW Cooling capacity (-40°C): 2,6 kW Heating capacity: 6 kW Pressure pump max.: 82 l/min / 1,5 bar Pump connection: M30x1,5 male Power supply: 400V 3~ 50Hz - w 1 * 1 items
Catalog # HUBE1052.0036.01
Supplier:  HUBER
New Product
Refrigerated Heating Circulator Temperature range: -60 ... 200 °C Cooling capacity (0°C): 7 kW Cooling capacity (-40°C): 2,6 kW Heating capacity: 6 kW Pressure pump max.: 82 l/min / 1,5 bar Pump connection: M30x1,5 male Power supply: 400V 3~ 50Hz - w 1 * 1 items
Catalog # HUBE1052.0036.01
Supplier:  HUBER