ContactUsLinkComponent
ActivArmr 47403 - Comfortable and flexible abrasion-resistant gloves, providing outstanding oil repellence - Nitrile coating: Superb abrasion and puncture resistance - Cotton lining: Optimal comfort and flexibility - Elasticated knitwrist: A snug fi 1 * 1 PAIR
Catalog # ANSE47-403/9
Supplier:  Ansell
New Product
ActivArmr 47403 - Comfortable and flexible abrasion-resistant gloves, providing outstanding oil repellence - Nitrile coating: Superb abrasion and puncture resistance - Cotton lining: Optimal comfort and flexibility - Elasticated knitwrist: A snug fi 1 * 1 PAIR
Catalog # ANSE47-403/9
Supplier:  Ansell