ContactUsLinkComponent
HPLC Waste Collector Justrite 6x UNF 1/4 , 7x NPT 1/8 , made of static-dissipative PE-EX, 6 capillary connections (UNF 1/4 ), 7 connections for tubing ID 5,5-8,0 mm (NPT1/8 ), coupling made of stainless steel. Accessories: Fittings made of PFA (6x gr 1 * 1 items
Catalog # BOHLM825-01
Supplier:  Bohlender
New Product
HPLC Waste Collector Justrite 6x UNF 1/4 , 7x NPT 1/8 , made of static-dissipative PE-EX, 6 capillary connections (UNF 1/4 ), 7 connections for tubing ID 5,5-8,0 mm (NPT1/8 ), coupling made of stainless steel. Accessories: Fittings made of PFA (6x gr 1 * 1 items
Catalog # BOHLM825-01
Supplier:  Bohlender