ContactUsLinkComponent
1/4 GC ferrule 1/4 ID (6.5mm) 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215465
New Product
1/4 GC ferrule 1/4 ID (6.5mm) 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215465