1/8 GC ferrule 0.8 mm ID two- hole 85% Vespel / 15% Graphite PK10 1 * 10 items
SGEA215288
New Product
1/8 GC ferrule 0.8 mm ID two- hole 85% Vespel / 15% Graphite PK10 1 * 10 items
SGEA215288
New Product
Frequently Bought Together
Avantor® ACE® C18-AR, HPLC Columns, 5 µm
: Avantor
VWR®, Manual Crimper and Decapper
: VWR Collection
EPA screw neck vials, N 24
: MACHEREY-NAGEL
VWR®, Snap Ring Vials ND11
: VWR Collection
HPLC columns, LiChrospher®
: Merck
Consumables for Shimadzu autosamplers
: Trajan Scientific and Medical
VWR®, Screw Neck Vials ND13
: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
: VWR Collection
Screw caps for screw neck vials, N 24
: MACHEREY-NAGEL
Avantor® ACE® C8, HPLC Columns, 5 µm
: Avantor
Autosampler syringes
: Trajan Scientific and Medical