1/16 GC ferrule 0.9 mm ID (long) 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215109
Supplier: Trajan Scientific and Medical
New Product
1/16 GC ferrule 0.9 mm ID (long) 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215109
Supplier: Trajan Scientific and Medical
New Product
Frequently Bought Together
Accessories for connection system GLS 80
Supplier: DWK Life Sciences
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
Safety funnels with lid, 'MARCO' V2.0
Supplier: S.C.A.T.
Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Exhaust filters, V3.0
Supplier: S.C.A.T.
Screw neck vials, N 9
Supplier: MACHEREY-NAGEL
VWR®, Snap Cap Vials
Supplier: VWR Collection
Avantor® Alltima, HPLC Columns, C18
Supplier: Avantor
Screw caps for screw neck vials, N 18 and N 20
Supplier: MACHEREY-NAGEL
Vydac 214TP C4, HPLC columns
Supplier: Avantor