AFS purifier I replacement cartridge (not for air or O2 use) 1 * 1 items
Catalog # SGEA202910
Supplier: Trajan Scientific and Medical
New Product
AFS purifier I replacement cartridge (not for air or O2 use) 1 * 1 items
Catalog # SGEA202910
Supplier: Trajan Scientific and Medical
New Product
Frequently Bought Together
OBJEKTIV SONY SELP18200 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
Accessories for Heracell Vios CR CO₂ incubators, CTS series, Thermo Scientific™
Supplier: Thermo Scientific
Chromatography tubing, FEP, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
with Multi-Axis-stand (min. additional need : Lens and Monitor) 1 * 1 items
Supplier: VISION ENGINEERING
Hybridisation oven, HL-2000 HybriLinker
Supplier: UVP ULTRA VIOLET PRODUCTS
CAMPDEN PELLET DISPENSER FOR 20 MG PELLETS 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
UV/Vis Calibration Standards
Supplier: HELLMA
VWR®, Eluent buffers for amino acid analyzer
Supplier: VWR Chemicals
KOMPAKTER, MASSENFLUSSMESSER AUS EDELSTAHL FÜR NICHTKORROSIVE GASE.NIEDRIGER DRUCKABFALL OPTION 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS