ContactUsLinkComponent
TEGERA Clean 5100, Disposable glove intended for cleanrooms of ISO class 5 (100). Well-fitting with extra length, textured and fing ertips and treated surface to that provide an enhanced grip. 1 * 50 PAIR
Catalog # EJEN5100-8
Supplier:  EJENDALS
New Product
TEGERA Clean 5100, Disposable glove intended for cleanrooms of ISO class 5 (100). Well-fitting with extra length, textured and fing ertips and treated surface to that provide an enhanced grip. 1 * 50 PAIR
Catalog # EJEN5100-8
Supplier:  EJENDALS