ContactUsLinkComponent
100 mm length x 2.1 mm ID ProteCol bio-inert UHPLC glass lined column 6-32 thread 0.5 µm frit 1 * 1 items
Catalog # SGEA08291336
New Product
100 mm length x 2.1 mm ID ProteCol bio-inert UHPLC glass lined column 6-32 thread 0.5 µm frit 1 * 1 items
Catalog # SGEA08291336