ContactUsLinkComponent
5 Ampoules de 1 mL ,, 15 components: Furaldehyde [CAS:98-01-1] 500mg/l Furfuryl alcohol [CAS:98-00-0] 1000mg/l Myrcene [CAS:123-35-3] 1000mg/l 4-Methylanisole [CAS:104-93-8] 1000mg/l Eucalyptol [CAS:470-82-6] 1000mg/l 2-Methylphenol [CAS:95 1 * 5 Ampoul
Catalog # CMPLRD1219111.5
New Product
5 Ampoules de 1 mL ,, 15 components: Furaldehyde [CAS:98-01-1] 500mg/l Furfuryl alcohol [CAS:98-00-0] 1000mg/l Myrcene [CAS:123-35-3] 1000mg/l 4-Methylanisole [CAS:104-93-8] 1000mg/l Eucalyptol [CAS:470-82-6] 1000mg/l 2-Methylphenol [CAS:95 1 * 5 Ampoul
Catalog # CMPLRD1219111.5