ContactUsLinkComponent
Muffle Furnace L9/13/B510 Tmax1300 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
Catalog # 455-3211
Supplier:  NABERTHERM
New Product
Muffle Furnace L9/13/B510 Tmax1300 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
Catalog # 455-3211
Supplier:  NABERTHERM