ContactUsLinkComponent
Muffle Furnace L5/13/C550 Tmax1300 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in f urnace door 1 * 1 items
Catalog # 455-3138
Supplier:  NABERTHERM
New Product
Muffle Furnace L5/13/C550 Tmax1300 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in f urnace door 1 * 1 items
Catalog # 455-3138
Supplier:  NABERTHERM