ContactUsLinkComponent
Muffle Furnace L3/12/C550 Tmax1200 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in f urnace door 1 * 1 items
Catalog # 455-2587
Supplier:  NABERTHERM
New Product
Muffle Furnace L3/12/C550 Tmax1200 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in f urnace door 1 * 1 items
Catalog # 455-2587
Supplier:  NABERTHERM