ContactUsLinkComponent
Ammonium standard solution (1 ppm NH4) 250 times concentrated Conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # ACSD5000302C
Supplier:  CPACHEM
New Product
Ammonium standard solution (1 ppm NH4) 250 times concentrated Conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # ACSD5000302C
Supplier:  CPACHEM