Accessories for Vios™ iDx CO₂ Incubators
390-1938
New Product
- Description:REINFORCEABLE SHELVES SS 160 - factory fitted option, replaces three standard shelves
- Seq:4
- Environmentally Preferable:
- For:Vios iDx 165, Vios iDx CR 165
Frequently Bought Together