ContactUsLinkComponent
Pump module VD 115 with vacuum pump VP1 - 230 V / 50-60 Hz version, pump cabinet for vacuum pumps, with switchable socket. Includes chemical diaphragm pump with nominal pumping speed of 2,0 m /h, final pressure of 7 mbar, and connection kit VD VP1-VP 1 * 1 items
Catalog # BIND8012-2624
Supplier:  Binder
New Product
Pump module VD 115 with vacuum pump VP1 - 230 V / 50-60 Hz version, pump cabinet for vacuum pumps, with switchable socket. Includes chemical diaphragm pump with nominal pumping speed of 2,0 m /h, final pressure of 7 mbar, and connection kit VD VP1-VP 1 * 1 items
Catalog # BIND8012-2624
Supplier:  Binder