ContactUsLinkComponent
1 component: C from Sodium Carbonate 1200mg/l CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # CMPLRM121774L1.50
New Product
1 component: C from Sodium Carbonate 1200mg/l CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # CMPLRM121774L1.50