ContactUsLinkComponent
Mounting kit with all necessary small parts to equip the vacuum pump VP1 or VP2 with an emission condenser at the pump outlet. The exhaust condenser can be operated with coolant and is used to recover the dried liquids as completely as possible. 1 * 1 KIT
Catalog # BIND8012-2615
Supplier:  Binder
New Product
Mounting kit with all necessary small parts to equip the vacuum pump VP1 or VP2 with an emission condenser at the pump outlet. The exhaust condenser can be operated with coolant and is used to recover the dried liquids as completely as possible. 1 * 1 KIT
Catalog # BIND8012-2615
Supplier:  Binder