ContactUsLinkComponent
Connection kit VD VP1-VP2 - for vacuum pump VP1 und VP2. Include a 1,9 m long plastic hose, clamping ring, universal centering ring and adapter small flange DN 16 to hose nozzle DN 10. 1 * 1 KIT
Catalog # BIND8012-2613
Supplier:  Binder
New Product
Connection kit VD VP1-VP2 - for vacuum pump VP1 und VP2. Include a 1,9 m long plastic hose, clamping ring, universal centering ring and adapter small flange DN 16 to hose nozzle DN 10. 1 * 1 KIT
Catalog # BIND8012-2613
Supplier:  Binder