ContactUsLinkComponent
Chlorides (Cl-) 2000mg/l in Water for Coulometric Titration Method CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C PACKAGING 1000 ML 1 * 1 L
Catalog # CMPLFC689649
New Product
Chlorides (Cl-) 2000mg/l in Water for Coulometric Titration Method CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C PACKAGING 1000 ML 1 * 1 L
Catalog # CMPLFC689649