ContactUsLinkComponent
Includes (4) clamps and (2) 5' Viton tubing (uninsulated), temperature range of -30°C to +200°C, 12mm ø 1 * 1 items
Catalog # HAKALT1600146
New Product
Includes (4) clamps and (2) 5' Viton tubing (uninsulated), temperature range of -30°C to +200°C, 12mm ø 1 * 1 items
Catalog # HAKALT1600146