ContactUsLinkComponent
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Heat Seal Closure, With Steam Indicator Dot, Size: 16 x 17 1 * 500 items
Catalog # KACC01617TWBBSD
New Product
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Heat Seal Closure, With Steam Indicator Dot, Size: 16 x 17 1 * 500 items
Catalog # KACC01617TWBBSD