ContactUsLinkComponent
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Self Seal Closure, With Steam Indicator Dot, Size: 16 x 35 1 * 250 items
Catalog # KACC01635TWBBSSSD
New Product
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Self Seal Closure, With Steam Indicator Dot, Size: 16 x 35 1 * 250 items
Catalog # KACC01635TWBBSSSD