ContactUsLinkComponent
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Heat Seal Closure, With Steam Indicator Dot, Size: 10 x 20 1 * 500 items
Catalog # KACC01020TWBBSD
New Product
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Heat Seal Closure, With Steam Indicator Dot, Size: 10 x 20 1 * 500 items
Catalog # KACC01020TWBBSD