ContactUsLinkComponent
Copper (Cu 2+) 10mg/l in Water CRM, ISO 17034 and ISO 17025,,,, Conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, packaging 100 ml bottle 1 * 100 mL
Catalog # CMPLRM039880L1
New Product
Copper (Cu 2+) 10mg/l in Water CRM, ISO 17034 and ISO 17025,,,, Conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, packaging 100 ml bottle 1 * 100 mL
Catalog # CMPLRM039880L1