ContactUsLinkComponent
1 component: Chromium (III) (Cr 3+) 10mg/l in Water CRM, ISO 17034 and ISO 17025,,,, Conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C PAKAGING 100 ML BOTTLE 1 * 100 mL
Catalog # CMPLRM019586L1
New Product
1 component: Chromium (III) (Cr 3+) 10mg/l in Water CRM, ISO 17034 and ISO 17025,,,, Conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C PAKAGING 100 ML BOTTLE 1 * 100 mL
Catalog # CMPLRM019586L1