ContactUsLinkComponent
100 barg pressure gauge with fittings and certification. This gauge is completely interchangeable with the existing gauge and can be used with or without the Raman cable support bracketry 1 * 1 items
Catalog # ASYNASY-C-1937
Supplier:  Asynt
New Product
100 barg pressure gauge with fittings and certification. This gauge is completely interchangeable with the existing gauge and can be used with or without the Raman cable support bracketry 1 * 1 items
Catalog # ASYNASY-C-1937
Supplier:  Asynt