ContactUsLinkComponent
Human GNLY/Granulysin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human GNLY/Granulysin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 96 Tests
Catalog # ANTIA2598-96
Supplier:  ANTIBODIES.COM
New Product
Human GNLY/Granulysin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human GNLY/Granulysin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 96 Tests
Catalog # ANTIA2598-96
Supplier:  ANTIBODIES.COM