ContactUsLinkComponent
Mouse Uteroglobin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse Uteroglobin in serum, plasma, tissue homogenates, cell lysates, lung lavage fluid, cell culture supern 1 * 96 Tests
Catalog # ANTIA2008-96
Anbieter:  Antibodies.com
New Product
Mouse Uteroglobin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse Uteroglobin in serum, plasma, tissue homogenates, cell lysates, lung lavage fluid, cell culture supern 1 * 96 Tests
Catalog # ANTIA2008-96
Anbieter:  Antibodies.com