ContactUsLinkComponent
Human PI 3 Kinase Class 2A/Cpk ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human PI 3 Kinase Class 2A/Cpk in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1878-96
Supplier:  ANTIBODIES.COM
New Product
Human PI 3 Kinase Class 2A/Cpk ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human PI 3 Kinase Class 2A/Cpk in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1878-96
Supplier:  ANTIBODIES.COM