ContactUsLinkComponent
Human T Plastin/PLS3 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of human T Plastin/PLS3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA270853-96
New Product
Human T Plastin/PLS3 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of human T Plastin/PLS3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA270853-96