ContactUsLinkComponent
Human SOD2/MnSOD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human SOD2/MnSOD in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA2179-96
Supplier:  ANTIBODIES.COM
New Product
Human SOD2/MnSOD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human SOD2/MnSOD in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA2179-96
Supplier:  ANTIBODIES.COM