ContactUsLinkComponent
Mouse Fas Ligand ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse Fas Ligand in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA1690-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Fas Ligand ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse Fas Ligand in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 96 Tests
Catalog # ANTIA1690-96
Supplier:  ANTIBODIES.COM